SLW1130/05M
Pyrex® Erlenmeyer flask, narrow neck, with printed trace code
capacity 10 mL
Synonym(s):
Pyrex, SciLabware 1130/05M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
flat bottom
Joint
Joints neck (12)
sterility
non-sterile
feature
graduated
without cap
printed trace code
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1130/05M
H
60 mm
body O.D.
30 mm
capacity
10 mL
neck I.D.
12 mm
fitting
narrow neck
Looking for similar products? Visit Product Comparison Guide
General description
- Complies with DIN ISO 1773
- Erlenmeyer pattern
- Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
- Graduated to full capacity
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
Features and Benefits
• Complies with DIN ISO 1773• Erlenmeyer pattern• Manufactured from PYREX® borosilicate glass for highest resistance to thermal shock and chemical corrosion• White enamel graduations and marking spot• Graduated to full capacity• Printed with Trace Code for downloadable batch certificate• 5, 10, 125, 150, 200, 300 and 5000 ml flasks are supplementary to the ISO range. 125 ml flask comforms to ASTM E1404 - 94.
Other Notes
Browse all Erlenmeyer flasks
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service