Skip to Content
Merck
All Photos(1)

Key Documents

P27135

Sigma-Aldrich

Phenylmercuric chloride

Synonym(s):

Phenylmercury chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5HgCl
CAS Number:
Molecular Weight:
313.15
Beilstein:
3536717
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

248-250 °C (dec.) (lit.)

SMILES string

Cl[Hg]c1ccccc1

InChI

1S/C6H5.ClH.Hg/c1-2-4-6-5-3-1;;/h1-5H;1H;/q;;+1/p-1

InChI key

AWGTVRDHKJQFAX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Sorsa et al.
Medical biology, 63(2), 66-72 (1985-01-01)
Latent and active collagenase were extracted from human polymorphonuclear leukocytes. Separation of the two forms of the enzyme was performed by gel filtration on Sepharose 6 B. The latent form of the enzyme was detected from chromatographic fractions after a
V Lemau de Talancé et al.
Bioorganic & medicinal chemistry letters, 21(24), 7265-7267 (2011-11-15)
This study describes two novel synthetic procedures to prepare APM, a useful tool for the analysis and the purification of thiolated biomolecules. The methods developed are technically simple and robust and allowed the first full characterization of pure APM. Moreover
Gelatinases/type IV collagenases in jaw cyst expansion.
O Teronen et al.
Annals of the New York Academy of Sciences, 732, 486-488 (1994-09-06)
James E Keller et al.
Neurochemical research, 28(3-4), 477-482 (2003-04-05)
Previous work had demonstrated that organomercurial-mediated modification of two cysteine residues in the vesicular acetylcholine transporter (VAChT) from Torpedo californica inhibits binding of vesamicol. The cysteines are protected by acetylcholine and vesamicol (Keller et al. 2000. J. Neurochem. 74:1739-1748). Modified
J Chmielnicka et al.
Ecotoxicology and environmental safety, 7(2), 165-171 (1983-04-01)
The effect of sodium selenite on the activity of the selected enzymes in blood serum and on mercury concentration in some tissues of guinea pigs exposed to ethyl- (EtHg) or phenylmercuric chloride (PhHg) was investigated. Every second day for a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service