Skip to Content
Merck
All Photos(1)

Key Documents

O601

Sigma-Aldrich

Octadecanamide

technical grade, 85%

Synonym(s):

Stearamide, Amide C18

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CONH2
CAS Number:
Molecular Weight:
283.49
Beilstein:
909006
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

form

powder

bp

250-251 °C/12 mmHg (lit.)

mp

98-102 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(N)=O

InChI

1S/C18H37NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H2,19,20)

InChI key

LYRFLYHAGKPMFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bo Yuan et al.
Journal of food and drug analysis, 26(2), 751-760 (2018-03-24)
African indigenous vegetables (AIVs) because of their nutrient density have the unique potential to reduce micronutrient deficiencies in sub-Saharan Africa, yet some may also contain anti-nutritive compounds. Vegetable nightshades from Solanum americanum, Solanum nigrum, Solanum scabrum and Solanum villosum are
S-S Li et al.
European review for medical and pharmacological sciences, 23(9), 3940-3950 (2019-05-23)
The aim of the study was to investigate the endogenous metabolites of patients with psoriasis vulgaris which will be helpful for the diagnosis of the disease and to provide the evidence of pathogenesis and the formulation for the individualized dosage
Pengchao Sun et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 86, 612-619 (2016-12-28)
Submicron emulsions (SEs) is an advanced formulation that possesses good biocompatibility, high loading of hydrophobic drugs, and good stability through autoclave sterilization. To enhance tumor targeting and tumor cell uptake, SEs could be modified with positive charge and targeting moieties.
Monika Zbucka-Kretowska et al.
Chemosphere, 203, 387-392 (2018-04-09)
Bisphenol A (BPA) is a synthetic chemical widely used in the industry, which may potentially evoke negative effects on human health, especially on reproductive processes and fetal development. BPA has been reported to act on estrogen, estrogen-related, androgen, thyroid hormone
Jidong Ru et al.
Scientific reports, 8(1), 13045-13045 (2018-08-31)
The main aim of this investigation was to promote the dyeing and level-dyeing effect of reactive dyes on cotton-fiber dyeing by encapsulating reactive dyes in liposomes as an alternative to sodium chloride. The results obtained indicated that liposomes, especially cationic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service