Skip to Content
Merck
All Photos(1)

Key Documents

MT51350056

Mettler-Toledo NIST/DIN buffer

pH 9.18

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€28.50

€28.50


Check Cart for Availability


Select a Size

Change View
250 ML
€28.50

About This Item

UNSPSC Code:
51191600
NACRES:
NB.79

€28.50


Check Cart for Availability

sterility

non-sterile

form

liquid

manufacturer/tradename

Mettler-Toledo 51350056

volume

250 mL

pH

9.18

Looking for similar products? Visit Product Comparison Guide

General description

Top Precision: buffers according to NIST and DIN 19266.
The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision. These buffers also come with a detailed test certificate and are only available in individual 250 mL bottles. Calibration cannot get better than this!

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service