Key Documents
MT51343150
Mettler Toledo InLab® Viscous & Viscous Pro pH electrode
Without temperature probe
Select a Size
1 EA
€518.00
€518.00
Check Cart for Availability
Select a Size
Change View
1 EA
€518.00
About This Item
UNSPSC Code:
41115604
NACRES:
NB.79
€518.00
Check Cart for Availability
material
glass membrane (polymer)
glass shaft (HA)
description
S7 connector
Without cable
manufacturer/tradename
Mettler-Toledo 51343150
L × W
40 mm × 6 mm
General description
The InLab® Viscous has been especially developed for viscous and highly glutinous samples.
The SteadyForce reference system guarantees the discharge of electrolytes, even with the most glutinous and fatty of samples, e.g. cosmetics, paints or resins.
The form has been chosen so that as little of the sample as possible adheres to the shaft and to facilitate cleaning. The result is a problem solver for most difficult sample types, the highly viscous materials.
  • high performance puncture electrode - especially designed for highly viscous
  • samples
  • easy to clean
  • SteadyForce reference system
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
SteadyForce is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)