Skip to Content
Merck
All Photos(1)

Key Documents

M2132

Sigma-Aldrich

mono-Methyl phosphate bis(cyclohexylammonium) salt

Synonym(s):

Phosphoric acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH5O4P · 2C6H13N
CAS Number:
Molecular Weight:
310.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

COP(O)(O)=O.NC1CCCCC1.NC2CCCCC2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service