Key Documents
HAM60675-01
Universal dispensing valve, for Hamilton® Microlab® 600 pipettor
right valve
Select a Size
1 EA
€369.00
€369.00
Available to ship on31 March 2025Details
Select a Size
Change View
1 EA
€369.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€369.00
Available to ship on31 March 2025Details
material
PTFE/CTFE
aluminum housing
feature
right valve
parameter
0-100 psig pressure
10-100 °C temperature (50-212 °F)
fitting
1/4-28 thread
suitability
suitable for (fits all Microlab 600 instruments)
General description
The Microlab 600 uses a universal valve that can support single dispensing, dual dispensing, diluting, and continuous dispensing. This valve functions on all Microlab 600 units. The valve has an aluminum housing with a PTFE-rotary plug. Syringes connect into the 5/16-32 threaded port.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)