H46600
3-Hydroxy-2-naphthoic hydrazide
97%
Synonym(s):
3-Hydroxy-2-naphthydrazide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC10H6CONHNH2
CAS Number:
Molecular Weight:
202.21
Beilstein:
395153
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
mp
205-208 °C (lit.)
SMILES string
NNC(=O)c1cc2ccccc2cc1O
InChI
1S/C11H10N2O2/c12-13-11(15)9-5-7-3-1-2-4-8(7)6-10(9)14/h1-6,14H,12H2,(H,13,15)
InChI key
FDNAQCWUERCJBK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service