Skip to Content
Merck
All Photos(1)

Key Documents

H1259

Sigma-Aldrich

1,1,2,2-Tetrakis(p-hydroxyphenyl)ethane

powder

Synonym(s):

1,1,2,2-Ethanetetra-p-phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22O4
CAS Number:
Molecular Weight:
398.45
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

powder

impurities

≤10% residual solvent

color

white to off-white

SMILES string

Oc1ccc(cc1)C(C(c2ccc(O)cc2)c3ccc(O)cc3)c4ccc(O)cc4

InChI

1S/C26H22O4/c27-21-9-1-17(2-10-21)25(18-3-11-22(28)12-4-18)26(19-5-13-23(29)14-6-19)20-7-15-24(30)16-8-20/h1-16,25-30H

InChI key

HDPBBNNDDQOWPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service