Skip to Content
Merck
All Photos(1)

Key Documents

H12404

Sigma-Aldrich

3-Hexanol

97%

Synonym(s):

Ethyl propyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH2CH3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718964
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.415 (lit.)

bp

134-135 °C (lit.)

density

0.820 g/mL at 20 °C
0.819 g/mL at 25 °C (lit.)

SMILES string

CCCC(O)CC

InChI

1S/C6H14O/c1-3-5-6(7)4-2/h6-7H,3-5H2,1-2H3

InChI key

ZOCHHNOQQHDWHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Belén Cotes et al.
Oecologia, 179(1), 151-162 (2015-05-07)
To locate and evaluate host patches before oviposition, parasitoids of herbivorous insects utilize plant volatiles and host-derived cues, but also evaluate predator-derived infochemicals to reduce predation risks. When foraging in host habitats infested with entomopathogenic fungi that can infect both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service