Skip to Content
Merck
All Photos(1)

Key Documents

GF63309810

Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8

500mm tube, 25.4mm outside diameter, 1.65mm wall thickness, 22.1mm inside diameter, annealed

Synonym(s):

NI027661

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 633-098-10

L × wall thickness

500 mm × 1.65 mm

O.D. × I.D.

25.4 mm × 22.1 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service