Skip to Content
Merck
All Photos(1)

Key Documents

GF49341247

Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti

500mm tube, 1.60mm outside diameter, 0.55mm wall thickness, 0.5mm inside diameter, hard

Synonym(s):

FE217160

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

Assay

99.9%

form

tubes

manufacturer/tradename

Goodfellow 493-412-47

L × wall thickness

500 mm × 0.55 mm

O.D. × I.D.

1.60 mm × 0.5 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service