F17601
5-Formylsalicylic acid
95%
Synonym(s):
2-Hydroxy-5-formylbenzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HCOC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
166.13
Beilstein:
2640881
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
mp
250 °C (dec.) (lit.)
SMILES string
OC(=O)c1cc(C=O)ccc1O
InChI
1S/C8H6O4/c9-4-5-1-2-7(10)6(3-5)8(11)12/h1-4,10H,(H,11,12)
InChI key
UTCFOFWMEPQCSR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Suja Shrestha et al.
Bioorganic & medicinal chemistry, 15(20), 6535-6548 (2007-08-19)
A series of compounds containing one or two salicylic acid moieties were synthesized, and their efficacy to inhibit the phosphohydrolase activity of PTP1B examined. Some of the methylenedisalicylic acid derivatives were potent inhibitors of PTP1B. Of those derivatives, 3c exhibited
Manabu Nakazono et al.
Clinica chimica acta; international journal of clinical chemistry, 436, 27-34 (2014-05-13)
Various styrylbenzene compounds were synthesized and evaluated as mainly Aβ amyloid sensors. These compounds, however, cannot be used for detecting amyloid deposition in peripheral nerves because of the inherent sensitivity of the compounds. These compounds often generate false positives especially
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service