Skip to Content
Merck
All Photos(2)

Documents

E47247

Sigma-Aldrich

Ethyl 2-pyridylacetate

98%

Synonym(s):

Ethyl 2-pyridineacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Beilstein:
123817
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.497 (lit.)

bp

70 °C/0.05 mmHg (lit.)

density

1.084 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)Cc1ccccn1

InChI

1S/C9H11NO2/c1-2-12-9(11)7-8-5-3-4-6-10-8/h3-6H,2,7H2,1H3

InChI key

IUDKTVXSXWAKJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit N Pandya et al.
Tetrahedron letters, 55(50), 6922-6924 (2014-12-17)
An efficient strategy for the synthesis of indolizines from readily available starting materials via oxidative C-H functionalization and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service