Skip to Content
Merck
All Photos(1)

Key Documents

E2269

Sigma-Aldrich

Sodium 1,2-ethanedisulfonate

≥98%

Synonym(s):

1,2-Ethanedisulfonic acid disodium salt, Disodium 1,2-ethanedisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO3SCH2CH2SO3Na
CAS Number:
Molecular Weight:
234.16
Beilstein:
3920126
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)CCS([O-])(=O)=O

InChI

1S/C2H6O6S2.2Na/c3-9(4,5)1-2-10(6,7)8;;/h1-2H2,(H,3,4,5)(H,6,7,8);;/q;2*+1/p-2

InChI key

BJTHMUJCKBTCFR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Compound for preparation of "edisylate" salts

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service