Skip to Content
Merck
All Photos(1)

Documents

DWKCPS1000P

Azlon Squat Form Cylinder with Molded Graduation with blue over print

polypropylene cylinder, class B, capacity 1000 mL , with +/10mL tolerance

Synonym(s):

CPS1000P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121805
NACRES:
NB.43

material

polypropylene cylinder

feature

class B
graduated

packaging

pkg of 1 ea

manufacturer/tradename

Azlon CPS1000P

capacity

1000 mL , with +/10mL tolerance

Looking for similar products? Visit Product Comparison Guide

Application

Offers increased stability especially when working in confined spaces

Features and Benefits

Unique self draining base to eliminate water collection during cleaning in laboratory washing machines

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service