Skip to Content
Merck
All Photos(1)

Key Documents

DWK218068654

Duran® Gl 45 Laboratory Glass Bottle

Synonym(s):

218068654

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€451.00

€451.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 EACH
€451.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

€451.00


Estimated to ship on01 June 2025


material

clear bottle
plastic-coated glass bottle (PU)

feature

with screw cap to drain contents: no (With pouring ring)

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 218068654

capacity

10000 mL

Application

Preparation of light-sensitive materials, Storage of light-sensitive materials, Transport and shipping of light-sensitive materials, Autoclaving of media and reagents, Culturing vessel, Boro 3.3, plastic coated

Features and Benefits

Standardised GL-thread and matching cap system for practicularly tight sealing and simple, clean pouring out; UV protection up to approx. 500 nm due to external colouration; The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties; plastic coatedVery durable and chemically resistant amber colour ; Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic print; Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service