Skip to Content
Merck
All Photos(1)

Key Documents

D111007

Sigma-Aldrich

2,2′-Dihydroxy-4,4′-dimethoxybenzophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3OC6H3(OH)]2CO
CAS Number:
Molecular Weight:
274.27
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

98%

mp

133-136 °C (lit.)

SMILES string

COc1ccc(c(O)c1)C(=O)c2ccc(OC)cc2O

InChI

1S/C15H14O5/c1-19-9-3-5-11(13(16)7-9)15(18)12-6-4-10(20-2)8-14(12)17/h3-8,16-17H,1-2H3

InChI key

SODJJEXAWOSSON-UHFFFAOYSA-N

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Cunha et al.
Environmental research, 143(Pt B), 65-71 (2015-05-20)
In the last decades, awareness regarding personal care products (PCP), i.e. synthetic organic chemicals frequently used in cosmetic and hygienic products, has become a forward-looking issue, due to their persistency in the environment and their potential multi-organ toxicity in both
Jian Li et al.
Analytical and bioanalytical chemistry, 407(20), 6137-6148 (2015-06-07)
Benzophenone-type UV filters (BPs) are ubiquitous in the environment. Transformation products (TPs) of BPs with suspected toxicity are likely to be produced during disinfection of water by chlorination. To quickly predict the toxicity of TPs, in this study, a novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service