CLS982620
Pyrex® culture tubes, screw cap with PTFE liner
O.D. × L 20 mm × 125 mm
Synonym(s):
Corning® 9826
About This Item
Recommended Products
material
borosilicate glass
cap (PTFE Liner)
round bottom tube
thread size
18-415
sterility
non-sterile
packaging
case of 192
pack of 96 ea
manufacturer/tradename
Corning 9826-20
O.D. × L
20 mm × 125 mm
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Pyrex® culture tubes are made of low-expansion borosilicate glass.
- Includes a phenolic screw cap to facilitate handling and sealing after autoclaving.
- The polytetrafluoroethylene (PTFE) faced glued-in rubber liner ensures a highly inert sealing face.
- White enamel marking spot for pencil notations.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service