CLS2105100
Pyrex® serialized/certified class A colored scale burettes, with PTFE stopcock plug
volume 100 mL, accuracy: 0.10 mL
Synonym(s):
burettes, glass burettes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
feature
subdivision 0.20 mL
manufacturer/tradename
Corning 2105-100
volume measuring range
100 mL, accuracy: 0.10 mL
volume
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
Corning® 2105
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service