Skip to Content
Merck
All Photos(1)

Documents

CDS008318

Sigma-Aldrich

(4S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)dione

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N2O5
CAS Number:
Molecular Weight:
392.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(O)cc15)[C@@](O)(CC)C(=O)OC4

InChI

1S/C22H20N2O5/c1-3-12-13-7-11(25)5-6-17(13)23-19-14(12)9-24-18(19)8-16-15(20(24)26)10-29-21(27)22(16,28)4-2/h5-8,25,28H,3-4,9-10H2,1-2H3/t22-/m0/s1

InChI key

FJHBVJOVLFPMQE-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B - STOT RE 1

Target Organs

Gastro-intestinal system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Céline Chu et al.
Fundamental & clinical pharmacology, 28(6), 652-660 (2014-03-05)
In a previous study, we showed that cetuximab, a monoclonal antibody directed towards epidermal growth factor receptor, could inhibit P-glycoprotein (P-gp), an efflux protein of ATP-binding cassette family, and lead to an increased P-gp substrate intracellular concentration. Cetuximab is given
Hiroaki Ozasa et al.
Cancer science, 105(8), 1032-1039 (2014-05-16)
The c-MET receptor tyrosine kinase is the receptor for hepatocyte growth factor. Recently, activation of the c-MET/hepatocyte growth factor signaling pathway was associated with poor prognosis in various solid tumors and was one of the mechanisms of acquired resistance to
Gabriela Gajek et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-09)
The incidence of gastrointestinal cancers is increasing every year. Irinotecan (CPT-11), a drug used in the treatment of colorectal cancer and gastric cancer, is metabolized by carboxylesterases to an active metabolite, SN-38, which is more cytotoxic. CAPE (caffeic acid phenethyl
Jian Yu et al.
Phytotherapy research : PTR, 28(10), 1577-1580 (2014-05-21)
The present study aims to investigate the influence of irinotecan's toxicity by the biotransformation of glucoaurantio-obtusin to aurantio-obtusin. Intraperitoneal administration (i.p.) of 100 mg/kg aurantio-obtusin significantly increased the toxicity of irinotecan, but the i.p. administration of 100 mg/kg glucoaurantio-obtusin showed
Lauriane Goldwirt et al.
Cancer chemotherapy and pharmacology, 74(1), 185-193 (2014-05-29)
Glioblastoma (GBM), the most common primary brain tumor in adults, is usually rapidly fatal with median survival duration of only 15 months and a 3-year survival rate of <7 %. Temozolomide (TMZ) is the only anticancer drug that has improved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service