Skip to Content
Merck
All Photos(1)

Key Documents

C96605

Sigma-Aldrich

Cyclodecanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H18(=O)
CAS Number:
Molecular Weight:
154.25
Beilstein:
1617127
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.482 (lit.)

bp

106-107 °C/12 mmHg (lit.)

mp

21-24 °C (lit.)

density

0.958 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCCCC1

InChI

1S/C10H18O/c11-10-8-6-4-2-1-3-5-7-9-10/h1-9H2

InChI key

SXOZDDAFVJANJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service