Skip to Content
Merck
All Photos(1)

Key Documents

C58703

Sigma-Aldrich

4-Chloro-3-nitrobenzaldehyde

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€77.00

€77.00

List Price€154.00Save 50%

Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€77.00

About This Item

Linear Formula:
ClC6H3(NO2)CHO
CAS Number:
Molecular Weight:
185.56
Beilstein:
778323
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€77.00

List Price€154.00Save 50%

Available to ship on03 April 2025Details


Request a Bulk Order

Assay

98%

form

powder

mp

61-63 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(C=O)ccc1Cl

InChI

1S/C7H4ClNO3/c8-6-2-1-5(4-10)3-7(6)9(11)12/h1-4H

InChI key

HETBKLHJEWXWBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alagarraj Arunadevi et al.
Nucleosides, nucleotides & nucleic acids, 38(9), 656-679 (2019-04-17)
In this work, we have synthesized a few novel mononuclear complexes of Cu(II), Co(II), Ni(II) and Zn(II) using a pyrazolone-derived Schiff base ligand. They were characterized by spectroscopic and analytical methods. The elemental analyses, UV-Vis, magnetic moment values and molar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service