Skip to Content
Merck
All Photos(1)

Key Documents

C58215

Sigma-Aldrich

4-Chloro-3-nitroaniline

97%

Synonym(s):

4-Amino-1-chloro-2-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)NH2
CAS Number:
Molecular Weight:
172.57
Beilstein:
1309394
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

powder

mp

99-101 °C (lit.)

SMILES string

Nc1ccc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C6H5ClN2O2/c7-5-2-1-4(8)3-6(5)9(10)11/h1-3H,8H2

InChI key

FOHHWGVAOVDVLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L O'Donoghue
Fundamental and applied toxicology : official journal of the Society of Toxicology, 6(3), 551-558 (1986-04-01)
4-Chloro-3-nitroaniline was given to groups of 20 male and 20 female rats by gavage at doses of 3.6, 18, or 90 mg/kg in a 10% corn oil suspension. Doses were administered 5 days per week for 90 days. The high
V Strauss et al.
Toxicology letters, 191(1), 88-95 (2009-08-18)
The impact of the strain on the metabolite profile of plasma samples in rats dosed with 2500 ppm 2-methyl-4-chlorophenoxyacetic acid (MCPA acid) or 45 mg/kg bw/day 4-chloro-3-nitroaniline (4C3N) for 4 weeks was evaluated. Four different strains were used: two Wistar
Sajad Jafari et al.
Journal of colloid and interface science, 490, 478-487 (2016-12-06)
Novel photocatalyts were synthesized by dispersion of AgX (X=I, Br, Cl) in nano clinoptilolite particles (NCP). The obtained composites (AgX-NCP) were used for the photodegrading of aqueous mixture of 4-methoxy aniline and 4-chloro-3-nitroaniline under Hg lamp irradiation. All samples were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service