Skip to Content
Merck
All Photos(1)

Documents

C011

Sigma-Aldrich

cis-2-Methyl-5-trimethylammoniummethyl-1,3-oxathiolane iodide

Synonym(s):

OXA-22

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18INOS
CAS Number:
Molecular Weight:
303.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

color

yellow

solubility

H2O: soluble
methanol: soluble

SMILES string

[I-].C[C@H]1O[C@H](CS1)C[N+](C)(C)C

InChI

1S/C8H18NOS.HI/c1-7-10-8(6-11-7)5-9(2,3)4;/h7-8H,5-6H2,1-4H3;1H/q+1;/p-1/t7-,8-;/m0./s1

InChI key

TWNAKQKXXMGYQT-WSZWBAFRSA-M

Biochem/physiol Actions

Potent muscarinic acetylcholine receptor agonist.

Caution

Photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Angeli et al.
British journal of pharmacology, 85(4), 783-786 (1985-08-01)
The efficacy and affinity of nine pentatomic cyclic muscarinic agonists were determined on the guinea-pig ileum, according to the method of Furchgott & Bursztyn (1967). The efficacy and affinity of these agonists are affected differently by structural modifications. Our results
R M Eglen et al.
Naunyn-Schmiedeberg's archives of pharmacology, 342(4), 394-399 (1990-10-01)
1. para-Fluoro-hexahydrosila-difenidol (p-F-HHSiD) has been proposed as an M3 selective antagonist. However, the M3 selectivity is variable in that it exhibits a high pA2 value for M3 muscarinic receptors in guinea-pig ileum but a low value at muscarinic receptors in
Zizhuo Su et al.
American journal of translational research, 12(7), 3964-3973 (2020-08-11)
Cyclophilin A (CyPA) plays an important role in the progression of atherosclerosis. Additionally, it has been reported that lysosomal function is markedly impaired in atherosclerosis induced by oxidized low-density lipoprotein (ox-LDL). As the CyPA degradation pathway remains to be elucidated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service