BR37412
BRAND® BLAUBRAND® volumetric flask
polypropylene stopper, amber borosilicate glass 3.3 flask, capacity 20 mL, accuracy, 0.06 mL
Synonym(s):
BRAND® BLAUBRAND® volumetric flask, PP stopper, amber glass
About This Item
Recommended Products
material
amber borosilicate glass 3.3 flask
polypropylene stopper
Joint
joint ST/NS12/21
description
calibrated to contain (TC, IN)
feature
accuracy 0.06 mL
class A DIN EN ISO 1042. Calibrated to contain (TC, In)
packaging
pack of 1 ea
manufacturer/tradename
BRAND 37412
capacity
20 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service