Skip to Content
Merck
All Photos(1)

Key Documents

BAF420741009

SP Bel-Art® Secador® 4.0 Gas-Purge Desiccator Cabinet

amber, volume 1.9 ft3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123002
NACRES:
NB.77
Pricing and availability is not currently available.

material

amber
polycarbonate

feature

Can be stacked three high

packaging

pkg of 1 ea

manufacturer/tradename

Bel-Art F42074-1009

W × D

34.1 cm (13.4 in.) × 41.4 cm (16.3 in.)

volume

1.9 ft3

General description

The attractive design and sturdy construction of the Secador® line of desiccator cabinets assures easy and reliable storage of moisture and UV sensitive products. Choose from an extensive selection of sizes, styles and colors to meet your specific dry storage needs. U.S. Patents: 6,834,920; 7,318,630 and D474,626

  • Polycarbonate construction blocks 99% of UV light* and is resistant to staining, crazing and chemical attack
  • Rear mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D.
  • Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front ; side swinging door opens to the left
  • Compact 34.1W x 41.4cmD footprint for space saving and can be stacked up to 3 units high
  • Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.
  • Gas ports can be closed to use desiccator with standard desiccant cartridges

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service