Key Documents
BAF420290001
SP Bel-Art® Techni-Dome® Polycarbonate Gas-Purge Desiccator
volume 2.3 ft3
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
material
clear polycarbonate
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F42029-0001
volume
2.3 ft3
General description
Generous quantities of standards, samples, electronics or other moisture-sensitive items can be protected in the Techni-Dome® 360 Vacuum Desiccator. A traditional style domed desiccator, the Techni-Dome® 360 is made from polycarbonate and offers a clear, unobstructed view of contents, with exceptionally large dry storage space, portability, and ease of use.
  • Spacious 65 liter (4000 cu. in.) internal volume can hold items up to 45cm (18”) tall or wide, including racks, trays and instruments
  • Two gas ports are tapered and accept tubing with ¼ to ⅜” I.D.; easy turning PTFE stopcocks facilitate turbulence-free entrance of gases, minimizing disturbance of contents
  • Polycarbonate construction is resistant to most common chemicals, as well as thermal stress; wipes clean easily
  • Includes two removable, polypropylene, 36.5cm(14.5”) and 22.5cm (9”) diameter shelves
  • Side handles provide easy portability and can be used as a place to affix tamper-evident seals
  • Gas ports can be closed to use desiccator with standard desiccant cartridges
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Techni-Dome is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)