Skip to Content
Merck
All Photos(2)

Key Documents

A22602

Sigma-Aldrich

2-Acetylthiophene

98%

Synonym(s):

Methyl-2-thienyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6OS
CAS Number:
Molecular Weight:
126.18
Beilstein:
107910
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.565 (lit.)

bp

214 °C (lit.)

mp

10-11 °C (lit.)

density

1.168 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cccs1

InChI

1S/C6H6OS/c1-5(7)6-3-2-4-8-6/h2-4H,1H3

InChI key

WYJOVVXUZNRJQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

206.6 °F

Flash Point(C)

97 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinbing Liu et al.
Bioorganic & medicinal chemistry, 16(3), 1096-1102 (2008-03-08)
A series of 1-(1-arylethylidene)thiosemicarbazide compounds and their analogues were synthesized and characterized by 1H NMR, MS. Their tyrosinase inhibitory activities were investigated by an assay based on the catalyzing ability of tyrosinase for the oxidation of L-DOPA, comparing with 4-methoxycinnamic
Mostafa M Ghorab et al.
Acta pharmaceutica (Zagreb, Croatia), 64(4), 419-431 (2014-12-23)
A novel series of thiophenes having biologically active sulfonamide 2-11, 3-methylisoxazole 12, 4-methoxybenzo[d] thiazole 13, quinoline 14, 15, benzoylphenylamino 16, and anthracene-9,10-dione 17 moieties were prepared. Structures of the newly synthesized compounds were established by elemental analysis and spectral data.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service