934984
Gold Microelectrode
diam. 10 μm
Synonym(s):
MF-2006
All Photos(1)
About This Item
diam.
10 μm
Related Categories
General description
Microelectrodes consist of small diameter metal wires or fibers (5-100 µm) sealed within tempered glass bodies. We offer gold microelectrodes (Working electrodes) with 10 µm diameter for R&D applications in electrochemistry.Dimensions of the electrode are approximately 7 cm long and 4 mm OD. The electrical connection is a 0.060" gold-plated pin. Each microelectrode is visually inspected and electrochemically tested before shipping.