Skip to Content
Merck
All Photos(1)

Documents

932817

Sigma-Aldrich

Tweezer Kit

Reverse Action, Stainless

Synonym(s):

Tweezer, Grippers, Tweezers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

Tweezer kit: Specially intended for applications where a highly precise self-closing action is required such as in microscopy and assembly applications. Swiss made reverse action tweezers are comfortable for soldering, insertion/extraction, and anti-wicking.

OAL: 125 mm. Width: M: 8 mm. Gap: N:1.5 mm. Tip length: P: 60 mm.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service