Skip to Content
Merck
All Photos(1)

Key Documents

927155

Sigma-Aldrich

DPEO

≥95%

Synonym(s):

N,N′-Bis(2-phenylethyl)ethanediamide

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€65.10
1 G
€165.00

€65.10


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
€65.10
1 G
€165.00

About This Item

Empirical Formula (Hill Notation):
C18H20N2O2
CAS Number:
Molecular Weight:
296.36
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

€65.10


Available to ship on15 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

SMILES string

O=C(C(NCCC1=CC=CC=C1)=O)NCCC2=CC=CC=C2

InChI

1S/C18H20N2O2/c21-17(19-13-11-15-7-3-1-4-8-15)18(22)20-14-12-16-9-5-2-6-10-16/h1-10H,11-14H2,(H,19,21)(H,20,22)

InChI key

QZXQKYMXZRGKLB-UHFFFAOYSA-N

Application

DPEO is a ligand for the copper-catalyzed coupling of aryl halides and alkyl alcohols.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhixiang Chen et al.
Journal of the American Chemical Society, 141(8), 3541-3549 (2019-01-29)
A robust and practical protocol for preparing alkyl aryl ethers has been developed, which relies on using two types of ligands to promote Cu-catalyzed alkoxylation of (hetero)aryl halides. The reaction scope is very general for a variety of coupling partners

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service