Skip to Content
Merck
All Photos(1)

Key Documents

908282

Sigma-Aldrich

5′-S-Methyl-13C-5′-thioadenosine-1′,2′,3′,4′,5′,-13C5

≥98 atom % 13C, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C5H15N5O3S
Molecular Weight:
303.29
MDL number:
UNSPSC Code:
41116105
Pricing and availability is not currently available.

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥95% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

[13CH3]S[13CH2][13C@@H]1[13C@@H](O)[13C@@H](O)[13C@H](N2C=NC3=C(N)N=CN=C32)O1

InChI key

WUUGFSXJNOTRMR-TWZNKHQZSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service