Skip to Content
Merck
All Photos(1)

Documents

907774

Sigma-Aldrich

Polyoxyethylene bis(azide)

Synonym(s):

O,O′-Bis(2-azidoethyl)polyethylene glycol, PEG-diazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N3CH2CH2(OCH2CH2)nN3
CAS Number:
UNSPSC Code:
12352200

form

viscous liquid

mol wt

PEG Mw ~400 Da

color

pale yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

PEG for cross-linking and used in ″click″ chemistry reactions. PEGylated materials have found broad use in drug delivery systems, virology, and immunology, as the incorporation of PEG improves pharmacological properties such as increased water solubility, enhanced resistance to degradation (protein hydrolysis), increased circulation half-life, and reduced antigenicity. Polymer for preparing enzyme conjugates soluble in organic solvents; promising drug carrier.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving the tensile strength of carbon nanotube yarn via one-step double [2+1] cycloadditions.
Kim H, et al.
Korean Journal of Chemical Engineering, 33(1), 299-304 (2016)
Shao Qiong Liu et al.
Biomaterials, 30(8), 1453-1461 (2008-12-23)
In this study, biodegradable PEG-peptide hydrogels have been synthesized using Click chemistry. A series of Arg-Gly-Asp (RGD) containing peptides were prepared via a solid phase synthesis approach, which were further functionalized with azide to yield peptide azide or peptide diazide.
Tilman Läppchen et al.
European journal of medicinal chemistry, 89, 279-295 (2014-12-03)
Calixarene 0118 is a potent anti-angiogenic agent that effectively inhibited tumor growth in preclinical studies, and is currently being evaluated in a phase I clinical trial. We have designed two close mimetics of calixarene 0118 containing a terminal alkynyl-functional group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service