Skip to Content
Merck
All Photos(1)

Key Documents

905828

Sigma-Aldrich

Dimethylsulfoxonium-3-(chloro)benzoylmethylide

≥95%

Synonym(s):

1-(3-Chlorophenyl)-2-(dimethyl(oxo)-sulfaneylidene)ethan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClO2S
CAS Number:
Molecular Weight:
230.71
UNSPSC Code:
12352101

Assay

≥95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

125-129 °C

functional group

chloro
ketone
sulfoxide

storage temp.

−20°C

SMILES string

O=C(C1=CC=CC(Cl)=C1)C=S(C)(C)=O

General description

Sulfoxonium ylides are highly stable carbene precursors, serving as a safer alternative to diazo compounds as they that do not generate gases as byproducts. Sulfoxonium ylides have been shown to be effective in a wide array of transition metal catalyzed C-H functionalization and coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaopeng Wu et al.
Organic letters, 20(5), 1396-1399 (2018-02-23)
A rhodium-catalyzed annulation between ethyl benzimidates and α- aroyl sulfur ylides was developed, affording a series of pyrano[4,3,2-ij]isoquinoline derivatives in moderate to good yields with good functional group compatibility. The procedure featured dual ortho-C-H functionalization and dual cyclization in one
James D Neuhaus et al.
Angewandte Chemie (International ed. in English), 57(49), 16215-16218 (2018-09-29)
A ruthenium-catalysed cross-olefination of diazo compounds and sulfoxonium ylides is presented. Our reaction design exploits the intrinsic difference in reactivity of diazo compounds and sulfoxonium ylides as both carbene precursors and nucleophiles, which results in a highly selective reaction.
Panjie Hu et al.
Organic letters, 20(8), 2160-2163 (2018-04-03)
Rh(III)-catalyzed cascade C-H activation of benzoylacetonitriles and annulation with sulfoxonium ylides was realized, leading to selective synthesis of naphthols, 2,3-dihydronaphtho[1,8- bc]pyrans, and naphtho[1,8- bc]pyrans. This step-economic reaction proceeded efficiently under mild and redox-neutral conditions via multiple C-H activations.

Related Content

Ketosulfoxonium ylides serve as versatile carbene equivalents for large-scale reactions, releasing dimethyl sulfoxide biproducts.

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service