Skip to Content
Merck
All Photos(1)

Key Documents

901590

Sigma-Aldrich

Phenol solution

1.0 M in THF

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

liquid

concentration

1.0 M in THF

refractive index

n/D 1.4222

density

0.922

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valuable building block offered as a solution in tetrahydrofuran for more convenient handling.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Nervous system,Kidney,Liver,Skin, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

2.0 °F

Flash Point(C)

-16.66 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service