Skip to Content
Merck
All Photos(1)

Documents

901228

Sigma-Aldrich

2-Ethylhexyl 4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate

Synonym(s):

2-Ethylhexyl-4,6-dibromo-3-fluorothieno[3,2-c]thiophene-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17Br2FO2S2
CAS Number:
Molecular Weight:
472.23
MDL number:
UNSPSC Code:
12352101

Assay

≥97%

form

powder

InChI

1S/C15H17Br2FO2S2/c1-3-5-6-8(4-2)7-20-15(19)12-10(18)9-11(21-12)14(17)22-13(9)16/h8H,3-7H2,1-2H3

InChI key

SOEJTOAKIMQIGL-UHFFFAOYSA-N

Application

2-Ethylhexyl 4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate (TTF) can be used in the synthesis of diketopyrrolopyrrole (DPP) based low band polymers for the fabrication of organic field-effect transistors (OFETs). It can also be used in the synthesis of narrow band-gap polymers like PBDTT-OS-TT-EF (1.52 eV) and PBDTT-OS-TT-CF (1.56 eV) for polymeric solar cells.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fine Tuning of Open-Circuit Voltage by Chlorination in Thieno[3,4-b]thiophene-Benzodithiophene Terpolymers toward Enhanced Solar Energy Conversion.
Qu S, et al.
Macromolecules, 50(13), 4962-4971 (2017)
PBDT-TSR: a highly efficient conjugated polymer for polymer solar cells with a regioregular structure.
Yao H, et al.
Journal of Material Chemistry A, 4(5), 1708-1713 (2016)
Toward high open-circuit voltage by smart chain engineering in 2D-conjugated polymer for polymer solar cells
Cheng X, et al.
Solar Energy Materials and Solar Cells, 149(82), 162-169 (2016)
An ultra-low bandgap diketopyrrolopyrrole (DPP)-based polymer with balanced ambipolar charge transport for organic field-effect transistors
Jiang T, et al.
Royal Society of Chemistry Advances, 6(82), 78720-78726 (2016)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service