Skip to Content
Merck
All Photos(1)

Key Documents

900197

Sigma-Aldrich

Reduced graphene oxide

greener alternative

stabilized with poly(sodium 4-styrenesulfonate), 10 mg/mL, dispersion in H2O

Synonym(s):

PSS-rGO, rGO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352119

form

dispersion in H2O

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10 mg/mL

greener alternative category

SMILES string

OC1=CC=C2C3=C1C(C=C(O)C4=C56)=C5C7=C8C9=C%10C%11=C%12C%13=C%14C%15=C%16C%17=C%18C(C(C(C%19=CC=C%20O)=C(C(O)=O)C=C%21%22)=C%21C%23=C%18C%24=C%16C%25=C%14C%26=C%12C%27=C%10C%28=C8C6=C%29=C%30C%28=C%31C%27=C%32C%26=C%33C%25=C%34C%24=C%35C%23=C%36C%22=CC(O)=C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". High concentrated graphene oxide sheets provide the prerequisite viscosity to bind the electrode materials together and enable 3D printing. Using water as a green solvent makes this aqueous ink system feasible for processing and drying safety and low cost. Click here for more information.

Application

  • Non-covalent functionalization with PSS molecules prevents rGO from aggregation.
  • Preparation for nanocomposite with water-dispersed graphene.
  • Electrochemical sensors with water-dispersed graphene.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huabin Yin et al.
Nanoscale research letters, 8(1), 422-422 (2013-10-19)
Aqueous solution synthesis of reduced graphene oxide-germanium nanoparticles (RGO-GeNPs) was developed using graphene oxide (GO) as stabilizer, which could be conducive to obtain better excellent electrical properties. The information about morphology and chemical composition of the nanomaterials were obtained by
Non-enzymatic detection of bilirubin based on a graphene-polystyrene sulfonate composite.
Balamurugan T, et al.
Royal Society of Chemistry Advances, 5(62), 50470-50477 (2015)
Influence of synthesis conditions on properties of green-reduced graphene oxide.
Pruna A, et al.
Journal of Nanoparticle Research, 15(5), 1605/1-1605/11 (2013)
Synthesis of poly(sodium 4-styrenesulfonate) functionalized graphene/cetyltrimethylammonium bromide (CTAB) nanocomposite and its application in electrochemical oxidation of 2,4-dichlorophenol.
Li J, et al.
Electrochimica Acta, 125, 1-8 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service