900020
Adiponitrile
battery grade, >99%, acid <200 ppm, H2O <100 ppm
Synonym(s):
1,4-Dicyanobutane
Select a Size
About This Item
Recommended Products
grade
battery grade
Quality Level
vapor density
3.7 (vs air)
vapor pressure
0.01 mmHg ( 40 °C)
Assay
>99%
form
liquid
autoignition temp.
1022 °F
expl. lim.
4.99 %
impurities
≤100 ppm H2O
≤200 ppm acid
refractive index
n20/D 1.438 (lit.)
bp
295 °C (lit.)
mp
1-3 °C (lit.)
density
0.951 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
N#CCCCCC#N
InChI
1S/C6H8N2/c7-5-3-1-2-4-6-8/h1-4H2
InChI key
BTGRAWJCKBQKAO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Caution
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.
Due to the adverse impact of the continued use of fossil fuels on the earth’s environment and climate, researchers have been asked to develop new approaches for producing power using renewable sources like wind and solar energy
Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service