Skip to Content
Merck
All Photos(1)

Key Documents

88544

Sigma-Aldrich

Thio-L-citrulline

≥98.0%

Synonym(s):

(S)-2-Amino-5-(thioureido)pentanoic acid, N5-Aminothioxomethyl-L-ornithine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N3O2S
CAS Number:
Molecular Weight:
191.25
Beilstein:
6844478
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

N[C@@H](CCCNC(N)=S)C(O)=O

InChI

1S/C6H13N3O2S/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)/t4-/m0/s1

InChI key

BKGWACHYAMTLAF-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent and selective inhibitor of neuronal and endothelial isoforms of nitric oxide synthase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Na Wu et al.
Pflugers Archiv : European journal of physiology, 472(12), 1743-1755 (2020-09-18)
Nitric oxide (NO) affects mitochondrial activity through its interactions with complexes. Here, we investigated regulations of complex I (C-I) and complex II (C-II) by neuronal NO synthase (nNOS) in the presence of fatty acid supplementation and the impact on left

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service