Skip to Content
Merck
All Photos(1)

Documents

860352

Sigma-Aldrich

2-Nitrophenyl β-D-thiogalactopyranoside

98%

Synonym(s):

2-Nitrophenyl 1-thio-β-D-galactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO7S
CAS Number:
Molecular Weight:
317.32
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

optical activity

[α]25/D −229.7°, c = 0.71 in methanol

mp

201 °C (dec.) (lit.)

SMILES string

OC[C@H]1O[C@@H](Sc2ccccc2[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

replaced by

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service