All Photos(1)
About This Item
Linear Formula:
H2NC6H4CH2CH(NH2)CO2H · xH2O
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352103
Assay
97%
mp
255-258 °C (dec.) (lit.)
SMILES string
O.NC(Cc1ccc(N)cc1)C(O)=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service