Skip to Content
Merck
All Photos(1)

Key Documents

82963

Sigma-Aldrich

2-(2-Pyridylazo)-1-naphthol

≥95.0%

Synonym(s):

ISOPAN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O
CAS Number:
Molecular Weight:
249.27
Beilstein:
480201
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0%

mp

124-128 °C

SMILES string

Oc1c(ccc2ccccc12)\N=N\c3ccccn3

InChI

1S/C15H11N3O/c19-15-12-6-2-1-5-11(12)8-9-13(15)17-18-14-7-3-4-10-16-14/h1-10,19H/b18-17+

InChI key

DKWJRBBUVAQOAC-ISLYRVAYSA-N

Other Notes

Reagent for the extraction and AAS determination of transition metals

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorota Wolicka et al.
Polish journal of microbiology, 64(2), 149-155 (2015-09-17)
Sulfate-reducing microbial communities were enriched from soils collected in areas with crude-oil exploitation. Cultures were grown in modified Postgate C medium and minimal medium, with ethanol or lactate as an electron donor. The batch cultures were grown with addition of
J. Komarek et al.
Collection of Czechoslovak Chemical Communications, 44, 3241-3241 (1979)
Kyle Lakatos et al.
Human vaccines & immunotherapeutics, 16(8), 1957-1968 (2020-01-30)
More effective rotavirus vaccines are essential for preventing extensive diarrheal morbidity and mortality in children under five years of age in low-resource regions. Nonreplicating rotavirus vaccines (NRRV) administered parenterally provide an alternate vaccination method to the current licensed oral vaccine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service