Skip to Content
Merck
All Photos(1)

Documents

805637

Sigma-Aldrich

XPhos-Pd-G2 GT capsule

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H59ClNPPd
Molecular Weight:
786.80
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

capsule

feature

generation 2

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

CC(C)C(C=C1C(C)C)=CC(C(C)C)=C1C2=CC=CC=C2C(C3CCCCC3)C4CCCCC4.NC5=C(C6=C([Pd]Cl)C=CC=C6)C=CC=C5

InChI

1S/C34H50.C12H10N.ClH.Pd/c1-23(2)28-21-31(24(3)4)34(32(22-28)25(5)6)30-20-14-13-19-29(30)33(26-15-9-7-10-16-26)27-17-11-8-12-18-27;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h13-14,19-27,33H,7-12,15-18H2,1-6H3;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

ZKKCVBQNSHPVBL-UHFFFAOYSA-M

Application

Convenient dosable form of XPhos-Pd-G2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The advent of the Buchwald portfolio of ligands and complexes for cross-coupling reactions in the past decade has greatly advanced the ease of performing this transformation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service