Skip to Content
Merck
All Photos(1)

Documents

804584

Sigma-Aldrich

(S,R)-Bode Kinetic Resolution Catalyst

Synonym(s):

(4aS,9aR)-6-Bromo-4-hydroxy-4,4a,9,9a-tetrahydroindeno[2,1-b][1,4]oxazin-3(2H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10BrNO3
CAS Number:
Molecular Weight:
284.11
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solution

storage temp.

15-25°C

SMILES string

O=C(CO1)N(O)[C@]2([H])[C@@]1([H])CC3=CC=C(Br)C=C32

InChI

1S/C11H10BrNO3/c12-7-2-1-6-3-9-11(8(6)4-7)13(15)10(14)5-16-9/h1-2,4,9,11,15H,3,5H2/t9-,11+/m1/s1

InChI key

FQXSUQCCAHKURK-KOLCDFICSA-N

Application

The following organocatalyst is used in the kinetic resolution of cyclic amines as reported by Bode and coworkers. In tandem with acylating reagent, (1E)-4-Hydroxy-1-mesityl-4-methylpent-1-en-3-one, either enantiomer can be accessed by using the catalyst shown or its enantiomer, both of which are available from Sigma-Aldrich.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Bode Group aims to develop new reactions and reagents for the synthesis of complex molecules. The Bode Group has developed N-mesityl-substituted NHCs as organocatalysts for the catalytic generation of reactive species including activated carboxylates, homoenolates, and enolates. These novel catalysts and reactions have made possible a new generation of highly enantioselective annulations from simple starting materials under mild reaction conditions, usually at room temperature and without added reagents. Furthering the goal of designing new reagents to enable the assembly of complex molecules, the Bode group has developed SnAP reagents for the facile, one-pot conversion of aldehydes into N-unprotected, saturated N-heterocycles, including bicyclic and spirocyclic structures. These easy to handle reagents provide a simple and robust alternative to the challenging and restrictive cross-coupling methods for the functionalization of saturated N-heterocycles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service