Skip to Content
Merck
All Photos(1)

Key Documents

795763

Sigma-Aldrich

{[3-(Trifluoromethyl)phenyl]methyl}phosphonic acid

Synonym(s):

3-CF3BPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8F3O3P
CAS Number:
Molecular Weight:
240.12
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

SMILES string

OP(CC1=CC(C(F)(F)F)=CC=C1)(O)=O

InChI

1S/C8H8F3O3P/c9-8(10,11)7-3-1-2-6(4-7)5-15(12,13)14/h1-4H,5H2,(H2,12,13,14)

InChI key

TVXYXFHWFHNWTJ-UHFFFAOYSA-N

Application

Tuning of surface energy, wetting and work function of metal oxides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quadrupolar donor?acceptor substituted oligo(phenylenevinylene)s?synthesis and solvatochromism of the fluorescence
Detert, H.;
Journal of Physical Organic Chemistry, 17, 1051-1056 (2004)
Peter J Hotchkiss et al.
Accounts of chemical research, 45(3), 337-346 (2011-10-21)
Transparent metal oxides, in particular, indium tin oxide (ITO), are critical transparent contact materials for applications in next-generation organic electronics, including organic light emitting diodes (OLEDs) and organic photovoltaics (OPVs). Understanding and controlling the surface properties of ITO allows for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service