Skip to Content
Merck
All Photos(1)

Key Documents

792411

Sigma-Aldrich

5,5′-Dibromo-4,4′-didodecyl-2,2′-bithiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H52Br2S2
CAS Number:
Molecular Weight:
660.69
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

56-60 °C

SMILES string

BrC(S1)=C(CCCCCCCCCCCC)C=C1C2=CC(CCCCCCCCCCCC)=C(Br)S2

InChI

1S/C32H52Br2S2/c1-3-5-7-9-11-13-15-17-19-21-23-27-25-29(35-31(27)33)30-26-28(32(34)36-30)24-22-20-18-16-14-12-10-8-6-4-2/h25-26H,3-24H2,1-2H3

InChI key

HTSYYLNTMSXYHZ-UHFFFAOYSA-N

Application

This monomer is used in the synthesis of semiconductive polymers for OPV and OFET devices

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Semiconductive Polymers Containing Dithieno[3; 2-b:2′; 3′-d]pyrrole for Organic Thin-Film Transistors
Wei Zhang, Jun Li, Li Zou, Bao Zhang, et al.
Macromolecules, 41(23), 8953-8955 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service