Skip to Content
Merck
All Photos(1)

Documents

791865

Sigma-Aldrich

1-(2-Bromoethoxy)-2-(2-methoxyethoxy)ethane

97%

Synonym(s):

1-(2-Bromoethoxy)-2-(2-methoxyethoxy)ethane, 1-bromo-3,6,9-trioxadecane, 2-[2-(2-methoxyethoxy)ethoxy]ethyl bromide, Diethylene glycol 2-bromoethyl methyl Ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15BrO3
CAS Number:
Molecular Weight:
227.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

liquid

refractive index

n/D 1.461

density

0.917 at 25 °C

storage temp.

2-8°C

SMILES string

BrCCOCCOCCOC

InChI

1S/C7H15BrO3/c1-9-4-5-11-7-6-10-3-2-8/h2-7H2,1H3

InChI key

LCCCTXULXHJDLA-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service