Skip to Content
Merck
All Photos(1)

Key Documents

773115

Sigma-Aldrich

Fmoc-N-Me-Met-OH

97% (HPLC)

Synonym(s):

N-Methyl methionine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4S
CAS Number:
Molecular Weight:
385.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

97% (HPLC)

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white to beige

mp

150-155 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CSCC[C@H](N(C)C(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H23NO4S/c1-22(19(20(23)24)11-12-27-2)21(25)26-13-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,18-19H,11-13H2,1-2H3,(H,23,24)/t19-/m0/s1

InChI key

SPYUJXKMEJUAOI-IBGZPJMESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service