Skip to Content
Merck
All Photos(1)

Key Documents

772607

Sigma-Aldrich

2-Fluoropyridine-4-carboxaldehyde

97%

Synonym(s):

2-Fluoroisonicotinaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€69.30

€69.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€69.30

About This Item

Empirical Formula (Hill Notation):
C6H4FNO
CAS Number:
Molecular Weight:
125.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€69.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.512

bp

357-403 °C (decomposition)

density

1.257 g/mL at 25 °C

functional group

aldehyde
fluoro

storage temp.

−20°C

SMILES string

Fc1cc(C=O)ccn1

InChI

1S/C6H4FNO/c7-6-3-5(4-9)1-2-8-6/h1-4H

InChI key

WOLIMEQXFWSBTE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

187.0 °F

Flash Point(C)

86.11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service